M8626
16-Methylene-17α-hydroxypregnenolone 3-acetate
Synonym(s):
3β,17α-Dihydroxy-16-methylene-5-pregnen-20-one acetate, 5-Pregnene-16-methylene-3β,17α-diol-20-one acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
SMILES string
[H][C@@]12CC=C3C[C@H](CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC(=C)[C@]4(O)C(C)=O)OC(C)=O
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.