Key Documents
M8557
Sigma-Aldrich
Myristic acid-carboxy-14C
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C14H22O11
Molecular Weight:
366.32
MDL number:
UNSPSC Code:
12352200
form
ethanol solution
extent of labeling
40-60 mCimmol
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
55.4 °F
flash_point_c
13 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.