Key Documents
M6042
Sigma-Aldrich
Anti-Macrophage Inflammatory Protein-1α antibody produced in goat
IgG fraction of antiserum, lyophilized powder
Synonym(s):
Anti-MIP-1α
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
51111800
biological source
goat
conjugate
unconjugated
antibody form
IgG fraction of antiserum
clone
polyclonal
form
lyophilized powder
species reactivity
human
technique(s)
indirect ELISA: suitable
western blot: suitable
storage temp.
−20°C
Gene Information
human ... CCL3(6348)
Specificity
The antibody will neutralize the biological activity of recombinant human, but not mouse, MIP-1α.
Immunogen
recombinant human MIP-1α expressed in E. coli
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline
Preparation Note
Purified using protein G.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)