Key Documents
M6005
Sigma-Aldrich
1-(trans-9-Octadecenoyl)-rac-glycerol
≥99%
Synonym(s):
Monoelaidin
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C21H40O4
CAS Number:
Molecular Weight:
356.54
MDL number:
UNSPSC Code:
12352211
assay
≥99%
storage temp.
−20°C
SMILES string
CCCCCCCC\C=C\CCCCCCCC(=O)OCC(O)CO
Choose from one of the most recent versions:
Certificates of Analysis (COA)