Key Documents
M5770
Sigma-Aldrich
8-Methylxanthine
~95% (TLC)
Synonym(s):
2,6-Dihydroxy-8-methylpurine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein/REAXYS Number:
171742
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
assay
~95% (TLC)
SMILES string
Cc1nc2NC(=O)NC(=O)c2[nH]1
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)