Key Documents
M5568
Sigma-Aldrich
MS-275
≥98% (HPLC), solid
Synonym(s):
3-pyridinylmethyl [[4-[[(2-aminophenyl)amino]carbonyl]phenyl]methyl]carbamate, N-(2-Aminophenyl)-4-[N-(pyridine-3ylmethoxycarbonyl)aminomethyl]benzamide
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C21H20N4O3
CAS Number:
Molecular Weight:
376.41
MDL number:
UNSPSC Code:
12352203
assay
≥98% (HPLC)
form
solid
solubility
DMSO: soluble 38 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
Nc1ccccc1NC(=O)c2ccc(CNC(=O)OCc3cccnc3)cc2
InChI
1S/C21H20N4O3/c22-18-5-1-2-6-19(18)25-20(26)17-9-7-15(8-10-17)13-24-21(27)28-14-16-4-3-11-23-12-16/h1-12H,13-14,22H2,(H,24,27)(H,25,26)
InChI key
INVTYAOGFAGBOE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
HDAC inhibitor; antiproliferative.
pictograms
Skull and crossbonesHealth hazard
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Repr. 1A
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
H Ottosson et al.
Scientific reports, 6, 36692-36692 (2016-11-09)
A new concept for treatment of infections is induction of our own antimicrobial peptides and the presented novel class of inducer, aroylated phenylenediamines (APDs), gives up to 20 to 30-fold induction of the human antimicrobial peptide LL-37, in vitro. In
Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the
France-Hélène Paradis et al.
Toxicological sciences : an official journal of the Society of Toxicology, 148(1), 220-228 (2015-08-08)
Histone deacetylases (HDACs) play a major role in chromatin remodeling, gene regulation, and cellular signaling. While the role of each class of HDAC during normal development is unclear, several HDAC inhibitors are embryotoxic; the mechanisms leading to the teratogenicity of