Key Documents
M4632
Supelco
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
Synonym(s):
2-Chlorodimedone
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H11ClO2
CAS Number:
Molecular Weight:
174.62
Beilstein/REAXYS Number:
2326749
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
161-164 °C (lit.)
format
neat
storage temp.
−20°C
SMILES string
CC1(C)CC(=O)C(Cl)C(=O)C1
Looking for similar products? Visit Product Comparison Guide
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Substrates
Substrate for chloroperoxidase.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
P Barnett et al.
The Journal of biological chemistry, 273(36), 23381-23387 (1998-08-29)
Here we describe the isolation, purification, and basic kinetic parameters of a vanadium type chloroperoxidase from the hyphomycete fungus Embellisia didymospora. The enzyme proved to possess similar high substrate affinities, a Km of 5 microM for a bromide, 1.2 mM
D Nowak et al.
Free radical biology & medicine, 16(4), 517-522 (1994-04-01)
We tested whether Ambroxol, a drug which stimulates the release of surfactant by pneumocytes type II, may also possess antioxidant properties. To assess the reactivity of Ambroxol with reactive oxygen species, we analysed its ability to decompose hydrogen peroxide (H2O2)
K W Zuurbier et al.
Biochimica et biophysica acta, 1037(2), 140-146 (1990-02-09)
The steady-state activity of myeloperoxidase in the chlorination of monochlorodimedone at neutral pH was investigated. Using a stopped-flow spectrophotometer we were able to show that the enzymic activity at pH 7.2 rapidly declined in time. During the first 50-100 ms
E de Boer et al.
The Journal of biological chemistry, 263(25), 12326-12332 (1988-09-05)
The reaction of vanadium-bromoperoxidase from the brown alga Ascophyllum nodosum with hydrogen peroxide, bromide, and 2-chlorodimedone has been subjected to an extensive steady-state kinetic analysis. Systematic variation of pH and the concentrations of these three components demonstrate that the reaction
Tina Kirchner et al.
Archives of biochemistry and biophysics, 495(1), 21-27 (2009-12-19)
The heme-containing enzyme myeloperoxidase (MPO) accumulates at inflammatory sites and is able to catalyse one- and two-electron oxidation reactions. Here it is shown that (-)-epicatechin, which is known to have numerous beneficial health effects, in low micromolar concentration enhances the