Key Documents
M3289
Sigma-Aldrich
Methyl 14-methylpalmitate
Synonym(s):
14-Methylhexadecanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H36O2
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352211
storage temp.
−20°C
SMILES string
CCC(C)CCCCCCCCCCCCC(=O)OC
Choose from one of the most recent versions:
Certificates of Analysis (COA)