Key Documents
M1521
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane
Synonym(s):
3-(Trimethoxysilyl)-1-propanethiol
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HS(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
196.34
Beilstein/REAXYS Number:
2038119
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form
liquid
refractive index
n20/D 1.444 (lit.)
bp
213-215 °C (lit.)
density
1.057 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CO[Si](CCCS)(OC)OC
InChI
1S/C6H16O3SSi/c1-7-11(8-2,9-3)6-4-5-10/h10H,4-6H2,1-3H3
InChI key
UUEWCQRISZBELL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
pictograms
Exclamation markEnvironment
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
204.8 °F - closed cup
flash_point_c
96 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Bioinspired functionalization of silica-encapsulated yeast cells.
Sung Ho Yang et al.
Angewandte Chemie (International ed. in English), 50(27), 6115-6118 (2011-06-01)
Galiya S Irmukhametova et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9551-9556 (2011-06-29)
A novel approach has been developed to synthesize thiolated sub-100 nm organosilica nanoparticles from 3-mercaptopropyltrimethoxysilane (MPTS) through its self-condensation in dimethylsulfoxide in contact with atmospheric oxygen. The formation of MPTS nanoparticles proceeds through the condensation of methoxysilane groups and simultaneous
Marcela Zanetti Corazza et al.
Journal of hazardous materials, 243, 326-333 (2012-11-20)
In the present study, the performance of multi-walled carbon nanotubes (MWCNTs) grafted with 3-mercaptopropyltrimethoxysilane (3-MPTMS), used as a solid phase extractor for Cd(2+) preconcentration in a flow injection system coupled to flame atomic absorption spectrometry (FAAS), was evaluated. The procedure
Eva Melnik et al.
Analytica chimica acta, 699(2), 206-215 (2011-06-28)
The process of surface functionalization involving silanization, biotinylation and streptavidin bonding as platform for biospecific ligand immobilization was optimized for thin film polyimide spin-coated silicon wafers, of which the polyimide film serves as a wave guiding layer in evanescent wave
Feng He et al.
ACS applied materials & interfaces, 4(8), 4373-4379 (2012-08-03)
The surfaces of Zn-doped biomagnetite nanostructured particles were functionalized with (3-mercaptopropyl)trimethoxysilane (MPTMS) and used as a high-capacity and collectable adsorbent for the removal of Hg(II) from water. Fourier transform infrared spectroscopy (FTIR) confirmed the attachment of MPTMS on the particle