Key Documents
M0251
Sigma-Aldrich
DL-Methionine methyl ester hydrochloride
95-97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C6H13NO2S · HCl
CAS Number:
Molecular Weight:
199.70
EC Number:
MDL number:
UNSPSC Code:
12352200
assay
95-97%
storage temp.
−20°C
SMILES string
Cl.COC(=O)C(N)CCSC
Choose from one of the most recent versions:
Certificates of Analysis (COA)