Key Documents
L8539
Sigma-Aldrich
L-687,384 hydrochloride
≥98% (HPLC), powder
Synonym(s):
1′-Benzyl-3,4-dihydrospiro­[naphthalene-1-(2H),4′-piperidine]
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C21H25N·HCl
Molecular Weight:
327.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Quality Level
assay
≥98% (HPLC)
form
powder
color
white
solubility
methanol: soluble 15 mg/mL at 60 °C
H2O: insoluble
SMILES string
Cl[H].C1Cc2ccccc2C3(C1)CCN(CC3)Cc4ccccc4
InChI
1S/C21H25N.ClH/c1-2-7-18(8-3-1)17-22-15-13-21(14-16-22)12-6-10-19-9-4-5-11-20(19)21;/h1-5,7-9,11H,6,10,12-17H2;1H
InChI key
ZKKYQWOBXVJKIR-UHFFFAOYSA-N
Gene Information
human ... OPRS1(10280)
Biochem/physiol Actions
σ1 receptor agonist.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters