Key Documents
L8391
Sigma-Aldrich
Anti-Luteinizing Hormone Releasing Hormone antibody produced in rabbit
whole antiserum, lyophilized powder
Synonym(s):
Anti-LHRH
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
whole antiserum
clone
polyclonal
form
lyophilized powder
species reactivity
human
technique(s)
radioimmunoassay: 1:200
storage temp.
2-8°C
Gene Information
human ... GNRH1(2796)
Specificity
The antibody is specific for LHRH and does not react with luteinizing hormone (LH), follicle stimulating hormone (FSH) or prolactin by RIA.
Immunogen
LHRH-BSA
Reconstitution
Reconstitute with 1 mL buffer: 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide. Store this stock at −20 °C. Working dilutions to be discarded if unused within 12 hours.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Federica Pimpinelli et al.
The European journal of neuroscience, 18(6), 1410-1418 (2003-09-27)
In this report we studied and compared the biochemical and the electrophysiological characteristics of two cell lines (GT1-7 and GN11) of immortalized mouse LHRH-expressing neurons and the correlation with their maturational stage and migratory activity. In fact, previous results indicated
F Pimpinelli et al.
Endocrinology, 140(1), 171-177 (1999-01-14)
PGs of the E series are involved in the control of LHRH secretion. The present experiments were conducted to clarify whether PGI2 (prostacyclin) might be also involved in such a control, using multiple methodological approaches on immortalized LHRH-secreting neurons. A