L7272
Methyl dodecanoate
≥99% (capillary GC)
Synonym(s):
Lauric acid methyl ester, Methyl laurate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)10COOCH3
CAS Number:
Molecular Weight:
214.34
Beilstein/REAXYS Number:
1767780
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
assay
≥99% (capillary GC)
bp
261-262 °C (lit.)
mp
4.5-5 °C
density
0.869 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCC(=O)OC
InChI
1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3
Inchi Key
UQDUPQYQJKYHQI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Replaced by
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.