Key Documents
L7262
Sigma-Aldrich
Lauroylcholine chloride
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C17H36NO2Cl
CAS Number:
Molecular Weight:
321.93
EC Number:
MDL number:
UNSPSC Code:
12352211
storage temp.
2-8°C
SMILES string
Cl.CCCCCCCCCCCC(=O)OCC[N](C)(C)C
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Eric H Hill et al.
ACS applied materials & interfaces, 7(9), 5550-5560 (2015-02-24)
Chemical and biological sensors are sought for their ability to detect enzymes as biomarkers for symptoms of various disorders, or the presence of chemical pollutants or poisons. p-Phenylene ethynylene oligomers with pendant charged groups have been recently shown to have