L6770
Lacto-N-tetraose
from human milk, ≥95% (HPAE/PAD)
Synonym(s):
β-D-Gal-(1→3)-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-D-Glc, LNT
About This Item
Recommended Products
biological source
human milk
assay
≥95% (HPAE/PAD)
storage temp.
2-8°C
SMILES string
CC(=O)N[C@H]1[C@@H](O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)O[C@H]3[C@@H](O)[C@@H](CO)O[C@@H](O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)[C@@H]3O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?