Key Documents
L0775
Sigma-Aldrich
1-Lauroyl-2-oleoyl-3-palmitoyl-rac-glycerol(C12:0/C18:1,[cis]-9/C16:0)
~98%
Synonym(s):
1-Dodecanoyl-2-(cis-9-octadecenoyl)-3-hexadecanoyl-rac-glycerol
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C49H92O6
CAS Number:
Molecular Weight:
777.25
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
assay
~98%
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.