MilliporeSigma
K1764
Sigma-Aldrich
Kazusamycin A
from Streptomyces sp., ≥95% (HPLC), aqueous methanol solution
Slide 1 of 1
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C33H48O7
CAS Number:
Molecular Weight:
556.73
MDL number:
UNSPSC Code:
12352200
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
biological source
Streptomyces sp.
assay
≥95% (HPLC)
form
aqueous methanol solution
solubility
methanol: water: soluble 7:3
DMSO: soluble
H2O: insoluble
chloroform: soluble
ethanol: soluble
hexane: insoluble
methanol: soluble
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCC(\C=C\C1OC(=O)C=CC1C)=C/C(C)C\C=C\C(C)=C\C(CO)C(=O)C(C)C(O)C(C)C\C(C)=C\C(O)=O
Biochem/physiol Actions
Kazusamycin A, an unsaturated branched chain fatty acid with a terminal lactone ring, is a hydroxy analog of Leptomycin B. It has significant in vitro cytotoxic activity against various human and mouse tumor lines encompassing a wide range of tissue types. Kazusamycin A exhibits in vivo antitumor activity against experimental murine tumors. It inhibits nuclear export and Rev translocation, a regulatory gene product in the HIV genome, at nanomolar concentrations.
Physical form
70% methanol solution.
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
60.0 °F - closed cup
flash_point_c
15.56 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
J B Tunac et al.
The Journal of antibiotics, 38(4), 460-465 (1985-04-01)
The novel, broad-spectrum antitumor antibiotics, CI-940 (PD 114,720) and PD 114,721 are components of bioactive compounds produced by a novel actinomycete (ATCC 39366). The organism is unique in that whole cell analysis revealed LL-diaminopimelic acid and major amounts of arabinose.
I Umezawa et al.
Gan to kagaku ryoho. Cancer & chemotherapy, 14(3 Pt 2), 858-864 (1987-03-01)
Kazusamycins A and B and leptomycin B have a structure characteristic of an unsaturated, branched-chain fatty acid with a terminal delta-lactone ring, and show antibacterial activity on some kinds of fungi. Kazusamycin A (KZM-A) showed cytotoxic activity on mammalian cells
Ryoichi Ando et al.
Bioorganic & medicinal chemistry letters, 16(12), 3315-3318 (2006-04-18)
Novel kazusamycin A derivatives were designed in the viewpoint of decrease of reactivity at the alpha,beta-unsaturated delta-lactone moiety against Michael-type addition. Although 25-30 steps were required for the synthesis of each compound, their syntheses were achieved. Cytotoxicity against HPAC cell
I Umezawa
Gan to kagaku ryoho. Cancer & chemotherapy, 11(12 Pt 2), 2674-2680 (1984-12-01)
The antitumor antibiotic sporamycin is composed of polypeptide and non-chromophore subunits and shows remarkable antitumor activity against various animal tumors. The mechanism of action of this compound involves host-mediated antitumor activity as well as direct cytotoxic activity due to the
The effect of kazusamycin on the growth of murine solid tumors and their spontaneous metastasis.
E Yoshida et al.
The Journal of antibiotics, 40(3), 391-393 (1987-03-01)