Key Documents
I8262
Sigma-Aldrich
Indole-3-acetic acid-carboxy-14C
~95% (Radiochemical Purity), aqueous ethanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
~95% (Radiochemical Purity)
form
aqueous ethanol solution
extent of labeling
1-15 mCi per mmol
mass shift
M+2
storage temp.
2-8°C
SMILES string
O[14C](=O)Cc1c[nH]c2ccccc12
Packaging
sealed ampule
Physical form
Acetone: ethanol (1:9) solution
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.