MilliporeSigma
All Photos(2)
Key Documents
I4382
Sigma-Aldrich
Ingenol 3,20-dibenzoate
≥99% (TLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C34H36O7
CAS Number:
Molecular Weight:
556.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
≥99% (TLC)
storage temp.
−20°C
SMILES string
C[C@@H]1C[C@@H]2[C@H]([C@H]3C=C(COC(=O)c4ccccc4)[C@@H](O)[C@@]5(O)[C@@H](OC(=O)c6ccccc6)C(C)=C[C@@]15C3=O)C2(C)C
Biochem/physiol Actions
Selectively activates the ε-isozyme of protein kinase C over the α- and β-isozymes.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)