Key Documents
I3271
Sigma-Aldrich
Interleukin-6 Fragment 88-121 human
≥80% (HPLC)
Synonym(s):
IL-6 (88-121)
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
51111800
biological source
human
assay
≥80% (HPLC)
form
solid
composition
Peptide content, ~70%
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... IL6(3569)
Biochem/physiol Actions
Competitively inhibits binding of IL-6 to its receptor.
Quality
Up to 20% of the methionine residue may be present as the sulfoxide.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
T Ekida et al.
Biochemical and biophysical research communications, 189(1), 211-220 (1992-11-30)
In order to locate the receptor-binding region of human interleukin-6 (IL-6), twelve peptide fragments were prepared by digestion of IL-6 with lysylendopeptidase. A significant activity of the receptor-binding was observed only for a peptide Ile88-Lys121, although the activity was estimated