Key Documents
I144
Sigma-Aldrich
L-chiro-Inositol 1,4,6-tris­phosphorothioate triethylammonium salt
solid
Sign Into View Organizational & Contract Pricing
Select a Size
Select a Size
Change View
About This Item
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
form
solid
color
light yellow-green
solubility
H2O: >10 mg/mL
methanol: >10 mg/mL
storage temp.
−20°C
SMILES string
CCN(CC)CC.O[C@H]1[C@H](O)[C@H](OP(O)(O)=S)[C@@H](OP(O)(O)=S)[C@@H](O)[C@@H]1OP(O)(O)=S
Biochem/physiol Actions
Selective inhibitor of myo-inositol.
Other Notes
The n value of the salt varies from 3 to 4 for each lot.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
P M Hansbro et al.
Biochemical and biophysical research communications, 200(1), 8-15 (1994-04-15)
The ability of the novel D-myo-inositol 1,4,5-trisphosphate [Ins(1,4,5)P 3] analogues, L-chiro-inositol 1,4,6-trisphosphate [L-chr Ins(1,4,6)P3] and the corresponding trisphosphorothioate compound [L-chr Ins(1,4,6)PS3] to inhibit soluble inositol (1,4,5)P3/(1,3,4,5)P4-polyphosphate 5-phosphatase, potently and selectively, has been investigated. L-chr Ins(1,4,6)P3 competitively inhibited 5-phosphate specific dephosphorylation
S T Safrany et al.
Biochemistry, 33(35), 10763-10769 (1994-09-06)
The interactions of synthetic analogues of D-myo-inositol 1,4,5-trisphosphate [Ins(1,4,5)P3] with the Ins(1,4,5)P3 receptor in permeabilized SH-SY5Y cells and with two key metabolic enzymes, Ins(1,4,5)P(3)3-kinase from a supernatant preparation of rat brain homogenates and Ins(1,4,5)P(3)5-phosphatase from human erythrocyte ghosts, have been
D Lampe et al.
Journal of medicinal chemistry, 37(7), 907-912 (1994-04-01)
Syntheses of novel phosphorothioate-based non-Ca(2+)-releasing inhibitors of myo-inositol 1,4,5-trisphosphate 5-phosphatase are described. 1L-myo-Inositol 1,4,5-trisphosphorothioate, myo-inositol 1,3,5-trisphosphorothioate, and 1L-chiro-inositol 1,4,6-trisphosphorothioate have been synthesized from 1L-2,3,6-tri-O-benzyl-1-O-(cis-prop-1-enyl)-myo-inositol, 2,4,6-tris-O-(p-methoxybenzyl)-myo-inositol orthoformate and 1L-2,3,5-tri-O-benzyl-chiro-inositol, respectively. 1L-2,3,5-Tri-O-benzyl-chiro-inositol was also used to prepare 1L-chiro-inositol 1,4,6-trisphosphate. The phosphorothioates did