Key Documents
H9770
Supelco
Hycanthone
~98%
Synonym(s):
1-([2-(Diethylamino)ethyl]amino)-4-(hydroxymethyl)-9H-thioxanthen-9-one
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C20H24N2O2S
CAS Number:
Molecular Weight:
356.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
assay
~98%
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
veterinary
storage temp.
−20°C
SMILES string
CCN(CC)CCNc1ccc(CO)c2Sc3ccccc3C(=O)c12
Gene Information
human ... CYP2D6(1565)
Choose from one of the most recent versions:
Certificates of Analysis (COA)