Key Documents
H7894
Sigma-Aldrich
gp120 Fragment 308-331
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C114H199N41O31
CAS Number:
Molecular Weight:
2640.06
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
composition
Peptide content, ~70%
UniProt accession no.
storage temp.
−20°C
Gene Information
human immunodeficiency virus 1 ... env(155971)
Amino Acid Sequence
Asn-Asn-Thr-Arg-Lys-Ser-Ile-Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys-Ile-Gly
Biochem/physiol Actions
Blocks fusion inhibition activity of both specific antiserum and HTLV-IIIβ-infected chimpanzee serum.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
J R Rusche et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(9), 3198-3202 (1988-05-01)
Antisera to recombinant human immunodeficiency virus (HIV) proteins containing the entire envelope, gp160, or the central portion of the envelope, PB1, can inhibit fusion of virally infected cells in culture. This fusion inhibition is HIV-variant specific--that is, anti-gp160-IIIB inhibits fusion