Key Documents
H6876
Sigma-Aldrich
2-Hydroxyhexanoic acid
95-98% (GC)
Synonym(s):
DL-α-Hydroxycaproic acid
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3(CH2)3CH(OH)CO2H
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
95-98% (GC)
mp
60-62 °C (lit.)
Storage temp.
−20°C
SMILES string
CCCCC(O)C(O)=O
InChI
1S/C6H12O3/c1-2-3-4-5(7)6(8)9/h5,7H,2-4H2,1H3,(H,8,9)
Inchi Key
NYHNVHGFPZAZGA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)