Key Documents
H4904
Sigma-Aldrich
(+)-Himbacine
~98% (GC), powder, muscarinic receptor antagonist
Synonym(s):
(3S,3aR,4R,4aS,8aR,9aS)-4-[(1E)-2-[(2R,6S)-1,6-dimethyl-2-piperdinyl]ethenyl]decahydro-3-methyl-naphtho[2,3-c]furan-1(3H)-one
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C22H35NO2
CAS Number:
Molecular Weight:
345.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
~98% (GC)
form
powder
optical activity
[α]/D +51.4°, c = 1.01 in chloroform(lit.)
color
off-white
solubility
ethanol: 50 mg/mL
H2O: insoluble
SMILES string
[H][C@]12CCCC[C@]1([H])[C@@H](\C=C\[C@@H]3CCC[C@@H](C)N3C)[C@@]4([H])[C@H](C)OC(=O)[C@@]4([H])C2
Gene Information
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Selective M2/M4 muscarinic acetylcholine receptor antagonist.
Caution
Hygroscopic
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
J H Miller et al.
The Journal of pharmacology and experimental therapeutics, 263(2), 663-667 (1992-11-01)
The binding potencies for the putative M2-selective antagonist himbacine were determined in radioligand binding and in functional response assays in neuronal tissue and Chinese hamster ovary cells containing transfected muscarinic receptors. Himbacine was shown to bind to all five cloned
Mariappan V Chelliah et al.
Journal of medicinal chemistry, 50(21), 5147-5160 (2007-09-15)
Pursuing our earlier efforts in the himbacine-based thrombin receptor antagonist area, we have synthesized a series of compounds that incorporate heteroatoms in the C-ring of the tricyclic motif. This effort has resulted in the identification of several potent heterocyclic analogs
Samuel Chackalamannil et al.
Journal of medicinal chemistry, 48(19), 5884-5887 (2005-09-16)
Structurally novel thrombin receptor (protease activated receptor 1, PAR-1) antagonists based on the natural product himbacine are described. The prototypical PAR-1 antagonist 55 showed a Ki of 2.7 nM in the binding assay, making it the most potent PAR-1 antagonist
Michaela Bitzer et al.
Experimental eye research, 82(3), 379-388 (2005-09-07)
Muscarinic antagonists, particularly atropine, can inhibit myopia development in several animal models and also in children. However, the biochemical basis of the inhibition of axial eye growth remains obscure, and there are doubts whether muscarinic receptors are involved at all.
Yoshito Takahashi et al.
Brain research, 1339, 26-40 (2010-04-14)
Cholinergic modulation of spontaneous GABAergic currents (mIPSC) was studied using whole-cell patch methods in mouse accessory olfactory bulb slices. Carbachol (above 100 microM) administration produced an increase in the mIPSC frequency in mitral cells, but did not affect the responses