Key Documents
H2774
Sigma-Aldrich
Hypercalcemia of malignancy factor fragment 107-139
≥90% (HPLC)
Synonym(s):
PTHrP (107-139), Parathyroid hormone-related protein (107-139), Thr-Arg-Ser-Ala-Trp-Leu-Asp-Ser-Gly-Val-Thr-Gly-Ser-Gly-Leu-Glu-Gly-Asp-His-Leu-Ser-Asp-Thr-Ser-Thr-Thr-Ser-Leu-Glu-Leu-Asp-Ser-Arg
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C142H228N42O58
CAS Number:
Molecular Weight:
3451.58
MDL number:
UNSPSC Code:
12352200
assay
≥90% (HPLC)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... PTHLH(5744)
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)