Key Documents
G9787
Sigma-Aldrich
3α,4β-Galactotriose
Synonym(s):
α-D-Gal-(1→3)-β-D-Gal-(1→4)-D-Gal, 4-O-(3-O-α-D-Galactopyranosyl-β-D-galactopyranosyl)-D-galactopyranose
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
UNSPSC Code:
12352201
storage temp.
−20°C
SMILES string
OC[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O[C@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H]1O)[C@H](O)[C@@H](O)C=O
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)