G9639
Controlled pore glass protein sequencing reagent
use tested, 200-400 mesh, pore size 75 Å
Synonym(s):
Controlled pore glass–uncoated, Glass, controlled-pore
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
particle size
200-400 mesh
pore size
75 Å
suitability
use tested
Application
Useful as an inert filler in solid phase protein sequencer columns.
Preparation Note
Acid washed
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)