Key Documents
G7264
Sigma-Aldrich
Big Gastrin I human
≥97% (HPLC)
Synonym(s):
Gregory Structure, HG-34, Human big gastrin I, pGlu-Leu-Gly-Pro-Gln-Gly-Pro-Pro-His-Leu-Val-Ala-Asp-Pro-Ser-Lys-Lys-Gln-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C176H251N43O53S
CAS Number:
Molecular Weight:
3849.20
EC Number:
MDL number:
UNSPSC Code:
12352200
assay
≥97% (HPLC)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... GAST(2520)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Storage form of gastrin that also has biological activity.
Antibody Adsorbant
H-pGlu-Leu-Gly-Pro-Gln-Gly-Pro-Pro-His-Leu-Val-Ala-Asp-Pro-Ser-Lys-Lys-Gln-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2 (+ xNH3)
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)