About This Item
Empirical Formula (Hill Notation):
C6H9N3O3 · HBr
CAS Number:
Molecular Weight:
252.07
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
Br.NCC(=O)NCC1=CC(=O)NO1
Choose from one of the most recent versions:
Certificates of Analysis (COA)