Key Documents
F1125
Sigma-Aldrich
Fast Red B Salt
Synonym(s):
2-Methoxy-4-nitrobenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo component 5
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C7H6N3O3 · 1/2ZnCl4
CAS Number:
Molecular Weight:
283.74
Colour Index Number:
37125
MDL number:
UNSPSC Code:
12171500
composition
Dye content, 15-20%
SMILES string
[Cl-].Cl[Zn]Cl.COc1cc(ccc1[NH+]=N)[N+]([O-])=O
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.