Key Documents
F0512
Sigma-Aldrich
(4-Formyl-3-methoxyphenoxy)acetic acid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H10O5
CAS Number:
Molecular Weight:
210.18
MDL number:
UNSPSC Code:
12352200
storage temp.
−20°C
SMILES string
COc1cc(OCC(O)=O)ccc1C=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)