Key Documents
E7394
Sigma-Aldrich
N-Acetyl-[D-Trp16]-Endothelin 1 fragment 16-21
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C46H62N8O10
CAS Number:
Molecular Weight:
887.03
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
composition
Dye content, ≥95% HPLC
UniProt accession no.
storage temp.
−20°C
SMILES string
[H][C@](NC(=O)[C@@]([H])(NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(C)=O)[C@@H](C)CC)([C@@H](C)CC)C(=O)N[C@@H](Cc3c[nH]c4ccccc34)C(O)=O
Gene Information
Amino Acid Sequence
Ac-D-Trp-Leu-Asp-Ile-Ile-Trp
Biochem/physiol Actions
ETA antagonist; inhibits ET-1 stimulated arachidonic acid release in rabbit vascular smooth muscle cells.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)