All Photos(1)
Documents
E7389
Sigma-Aldrich
cis-9:10-Epoxy-12-hydroxystearic acid methyl ester Mixture of isomers
~97% (capillary GC)
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C19H36O4
CAS Number:
Molecular Weight:
328.49
MDL number:
UNSPSC Code:
12352211
assay
~97% (capillary GC)
storage temp.
−20°C
SMILES string
CCCCCCC(O)CC1OC1CCCCCCCC(=O)OC
Packaging
Sealed ampule under argon
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.