E6253
Estriol 17β-(β-D-glucuronide)
Synonym(s):
1,3,5(10)-Estratriene-3,16α,17β-trihydroxy 17-glucuronide, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 17-glucuronide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H32O9
CAS Number:
Molecular Weight:
464.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC12CCC3C(CCc4cc(O)ccc34)C1CC(O)C2OC5OC(C(O)C(O)C5O)C(O)=O
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K Shimada et al.
Journal of chromatography, 232(1), 13-18 (1982-10-08)
A high-performance liquid chromatographic method for the determination of estriol 16-glucuronide and 17-glucuronide in bile and urine has been developed. The electrochemical detector was found to be more superior than the UV detector with respect to selectivity and sensitivity and