Key Documents
E4014
Sigma-Aldrich
Ethylenediamine-1,2-14C dihydrochloride
aqueous ethanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
H2N14CH214CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
133.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
aqueous ethanol solution
extent of labeling
1-10 mCi per mL
storage temp.
2-8°C
SMILES string
Cl.N[14CH2][14CH2]N
Looking for similar products? Visit Product Comparison Guide
Packaging
Sealed ampule
Physical form
Aqueous solution containing 2% ethanol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.