Key Documents
E2388
Sigma-Aldrich
Ethanolamine-1,2-14C hydrochloride
aqueous solution
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HO[14CH2]2NH2· HCl
CAS Number:
Molecular Weight:
97.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
aqueous solution
extent of labeling
5-25 mCi per mmol
mass shift
M+4
storage temp.
2-8°C
SMILES string
Cl.N[14CH2][14CH2]O
Packaging
Serum bottle
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.