E1520
Anti-β-Endorphin antibody produced in rabbit
whole antiserum
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
whole antiserum
clone
polyclonal
contains
15 mM sodium azide
species reactivity
rat
technique(s)
dot blot: 1:8,000 using β-endorphin (rat)
immunohistochemistry (frozen sections): 1:1,000 using 4% paraformaldehyde perfusion-fixed, frozen sections of rat pituitary
radioimmunoassay: 1:100 using 125I-β-endorphin (rat)
shipped in
dry ice
storage temp.
−20°C
Gene Information
rat ... Pomc(24664)
Immunogen
Choose from one of the most recent versions:
Certificates of Analysis (COA)