E0903
Ephrin-B3/Fc Chimera human
>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder
Synonym(s):
EFL-6, ELF-3, Elk-L3, LERK-8, NLERK-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf21 cells
assay
>95% (SDS-PAGE)
form
lyophilized powder
mol wt
monomer calculated mol wt 49.2 kDa
~58 kDa by SDS-PAGE (reducing)
storage condition
avoid repeated freeze/thaw cycles
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... EFNB3(1949)
Biochem/physiol Actions
Member of ephrin ligand family that binds Eph receptor tyrosine kinases (EphA4, EphB1, EphB2, and EphB3). Involved in pattern formation and morphogenesis.
Other Notes
Extracellular domain of human ephrin-B3 (amino acids 1-224) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.
Analysis Note
The biological activity is measured by its ability to bind immobilized recombinant mouse EphB3/Fc in ELISA.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?