Key Documents
D9879
Sigma-Aldrich
DNP-L-isoleucine
Synonym(s):
N-(2,4-Dinitrophenyl)-L-isoleucine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H15N3O6
CAS Number:
Molecular Weight:
297.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CCC(C)C(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)