Key Documents
D9512
Sigma-Aldrich
Didansylhistamine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C29H31N5O4S2
CAS Number:
Molecular Weight:
577.72
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
2-8°C
SMILES string
CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCc3cn(cn3)S(=O)(=O)c4cccc5c(cccc45)N(C)C
Choose from one of the most recent versions:
Certificates of Analysis (COA)