Key Documents
D8881
Sigma-Aldrich
Dansyl-DL-leucine cyclohexylammonium salt
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H24N2O4S · C6H13N
CAS Number:
Molecular Weight:
463.63
UNSPSC Code:
12352200
storage temp.
−20°C
SMILES string
NC1CCCCC1.CC(C)CC(NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)