Key Documents
D8415
Sigma-Aldrich
2′-Deoxyguanosine-8-14C
≥95% (Radiochemical Purity, HPLC), aqueous ethanol solution
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
≥95% (Radiochemical Purity, HPLC)
form
aqueous ethanol solution
extent of labeling
40-60 mCi per mmol
storage temp.
−20°C
SMILES string
NC1=NC(=O)c2ncn(C3CC(O)C(CO)O3)c2N1
Packaging
serum bottle
Physical form
Ethanol: water (1:1) solution
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.