Key Documents
D7314
Sigma-Aldrich
Deoxypeganine hydrochloride dihydrate
≥99% (HPLC), solid
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C11H12N2 · HCl · 2H2O
Molecular Weight:
244.72
MDL number:
UNSPSC Code:
12352200
assay
≥99% (HPLC)
form
solid
storage temp.
2-8°C
SMILES string
O.O.Cl.C1CN2Cc3ccccc3N=C2C1
Biochem/physiol Actions
Inhibits cholinesterases reversibly.
Caution
Light sensitive.
pictograms
Skull and crossbones
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)