Key Documents
D7254
Sigma-Aldrich
DNP-L-alanine
Synonym(s):
N-(2,4-Dinitrophenyl)-L-alanine
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9H9N3O6
CAS Number:
Molecular Weight:
255.18
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CC(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
C A Sotriffer et al.
Biophysical journal, 76(6), 2966-2977 (1999-06-04)
Antibody IgE Lb4 interacts favorably with a large number of different compounds. To improve the current understanding of the structural basis of this vast cross-reactivity, the binding of three dinitrophenyl (DNP) amino acids (DNP-alanine, DNP-glycine, and DNP-serine) is investigated in
P Margot et al.
Analytical biochemistry, 198(1), 15-18 (1991-10-01)
A sensitive and highly reproducible assay for N-acetylmuramoyl-L-alanine amidase (EC 3.5.1.28) was devised, based on specific and homogeneous L-[14C]alanine labeling of the substrate, the peptidoglycan. The method involves partial purification of both the enzyme and the substrate and monitoring the
H Nakayama et al.
European journal of biochemistry, 112(2), 403-409 (1980-11-01)
p-Amidinophenyl esters of an enantiomeric pair of N-(2,4-dinitrophenyl)alanine (N2Ph-Ala) were both efficiently hydrolyzed by trypsin. The acylation and deacylation rate constants for the D-isomer are 1/2.5 of those for the L-isomer. Slow rates of deacylation of the two substrates made