MilliporeSigma
All Photos(3)
Key Documents
D5422
Sigma-Aldrich
3α,6α-Mannotriose
≥95% (HPLC)
Synonym(s):
α-D-Man-(1→3)-[α-D-Man-(1→6)]-D-Man, 3,6-Di-O-(α-D-mannopyranosyl)-D-mannopyranose
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
assay
≥95% (HPLC)
storage temp.
−20°C
SMILES string
OCC1OC(OCC2OC(O)C(O)C(OC3OC(CO)C(O)C(O)C3O)C2O)C(O)C(O)C1O
Other Notes
Mannotriose core structure of N-linked oligosaccharides
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)