Key Documents
D2908
Sigma-Aldrich
2,6-Dichloroindophenol sodium salt hydrate
ACS reagent
Synonym(s):
2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt, 2,6-Dichlorobenzenone-indophenol sodium salt, 2,6-Dichlorophenolindophenol sodium salt hydrate, DCIP, DPIP
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H6Cl2NNaO2 · xH2O
CAS Number:
Molecular Weight:
290.08 (anhydrous basis)
Beilstein/REAXYS Number:
3641229
EC Number:
MDL number:
UNSPSC Code:
12171500
grade
ACS reagent
impurities
Interfering dyes, passes test
loss
≤12.0% loss on drying, 120°C
SMILES string
O.[Na+].[O-]c1ccc(cc1)\N=C2/C=C(Cl)C(=O)C(Cl)=C2
InChI
1S/C12H7Cl2NO2.Na.H2O/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7;;/h1-6,16H;;1H2/q;+1;/p-1
InChI key
XHSOLXWCGCVQHE-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)