Key Documents
D149
Sigma-Aldrich
Dihydro-β-erythroidine hydrobromide
solid
Synonym(s):
3β-1,6-Didehydro-14,17-dihydro-3-methoxy-16(15H)-oxaerythrinan-15-one hydrobromide
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C16H21NO3 · HBr
CAS Number:
Molecular Weight:
356.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
solid
optical activity
[α]22/D +99.2°, c = 0.90 in H2O(lit.)
color
white
solubility
ethanol: 5 mg/mL
H2O: 800 mg/mL
SMILES string
Br.CO[C@H]1CC=C2CC[N@@H]3CCC4=C(CC(=O)OC4)[C@]23C1
InChI
1S/C16H21NO3.BrH/c1-19-13-3-2-12-5-7-17-6-4-11-10-20-15(18)8-14(11)16(12,17)9-13;/h2,13H,3-10H2,1H3;1H/t13-,16-;/m0./s1
InChI key
GFIGWAJEIMHJJB-LINSIKMZSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Competitive nicotinic receptor antagonist.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Y T Wang et al.
Neuroscience, 40(3), 759-767 (1991-01-01)
The purpose of this study was to determine if ambigual oesophageal motoneurons of the rat possess functional nicotinic cholinoceptors. In urethane anaesthetized rats, acetylcholine (20-50 pmol) delivered micropneumophoretically from multibarrelled pipettes to the compact formation of the nucleus ambiguus produced
C Rapier et al.
Journal of neurochemistry, 54(3), 937-945 (1990-03-01)
Presynaptic nicotinic acetylcholine receptors on striatal nerve terminals modulate the release of dopamine. We have compared the effects of a number of nicotinic agonists and antagonists on a perfused synaptosome preparation preloaded with [3H]dopamine. (-)-Nicotine, acetylcholine, and the nicotinic agonists
Sean H White et al.
Journal of neurophysiology, 112(2), 446-462 (2014-04-18)
Nicotinic receptors form a diverse group of ligand-gated ionotropic receptors with roles in both synaptic transmission and the control of excitability. In the bag cell neurons of Aplysia, acetylcholine activates an ionotropic receptor, which passes inward current to produce a