Key Documents
D1256
Sigma-Aldrich
2′-Deoxyadenosine 3′:5′-cyclic monophosphate sodium salt
Synonym(s):
2′-deoxy-3′:5′-cAMP
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C10H11N5O5PNa
CAS Number:
Molecular Weight:
335.19
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Storage temp.
−20°C
SMILES string
[Na].Nc1ncnc2n(cnc12)C3CC4OP(O)(=O)OCC4O3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)