Key Documents
D0640
Sigma-Aldrich
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose
≥97%
Synonym(s):
D-Glucose diacetonide, Diacetone-D-glucose
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
Beilstein/REAXYS Number:
84386
EC Number:
MDL number:
UNSPSC Code:
12352200
assay
≥97%
mp
110-111 °C (lit.)
storage temp.
−20°C
SMILES string
CC1(C)OC[C@@H](O1)[C@H]2O[C@@H]3OC(C)(C)O[C@@H]3[C@H]2O
InChI
1S/C12H20O6/c1-11(2)14-5-6(16-11)8-7(13)9-10(15-8)18-12(3,4)17-9/h6-10,13H,5H2,1-4H3/t6-,7+,8-,9-,10-/m1/s1
InChI key
KEJGAYKWRDILTF-JDDHQFAOSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)